missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydrazinopyridine, 98%, ACROS Organics™

$35.34 - $35.34

Chemical Identifiers

CAS 4930-98-7
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006249
InChI Key NWELCUKYUCBVKK-UHFFFAOYSA-N
Synonym 2 1h-pyridinone, hydrazone, 2-hydrazinopyridine, 2-hydrazinylpyridine, 2-pyridylhydrazine, 2-pyridylhydrazone, hydrazine, 2-pyridinyl, pyridin-2-yl hydrazine, pyridin-2-yl-hydrazine, pyridine, 2-hydrazino, pyridine, 2-hydrazinyl
PubChem CID 78645
IUPAC Name pyridin-2-ylhydrazine
SMILES NNC1=CC=CC=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120870010
SDS View Documents
Thermo Fisher Scientific
AC120870010
1g Glass bottle
Each for $35.34
 
AC120870050
SDS View Documents
Thermo Fisher Scientific
AC120870050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC120870250
SDS View Documents
Thermo Fisher Scientific
AC120870250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4930-98-7
109.13
NWELCUKYUCBVKK-UHFFFAOYSA-N
78645
NNC1=CC=CC=N1
C5H7N3
MFCD00006249
2 1h-pyridinone, hydrazone, 2-hydrazinopyridine, 2-hydrazinylpyridine, 2-pyridylhydrazine, 2-pyridylhydrazone, hydrazine, 2-pyridinyl, pyridin-2-yl hydrazine, pyridin-2-yl-hydrazine, pyridine, 2-hydrazino, pyridine, 2-hydrazinyl
pyridin-2-ylhydrazine

Specifications

110°C
Glass bottle
Beige to White
1g
97.5
97.5% min. (GC)
MFCD00006249
2 1h-pyridinone, hydrazone, 2-hydrazinopyridine, 2-hydrazinylpyridine, 2-pyridylhydrazine, 2-pyridylhydrazone, hydrazine, 2-pyridinyl, pyridin-2-yl hydrazine, pyridin-2-yl-hydrazine, pyridine, 2-hydrazino, pyridine, 2-hydrazinyl
NNC1=CC=CC=N1
109.13
109.13
98%
Authentic
185.0°C (140.0 mmHg)
44.5°C to 46.0°C
4930-98-7
100.0
C5H7N3
22, I, 688
NWELCUKYUCBVKK-UHFFFAOYSA-N
pyridin-2-ylhydrazine
78645
Low Melting Solid
2-Hydrazinopyridine
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 225-566-8

SDS
Product Certifications