Learn More
2-Hydrazinopyrazine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6408606
Description
Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2-Hydrazinopyrazine | |
109°C to 113°C | |
C4H6N4 | |
5 g | |
2-hydrazinylpyrazine, 2-hydrazinopyrazine, pyrazin-2-yl-hydrazine, 1-pyrazin-2-yl hydrazine, hydrazinopyrazine, 2-pyrazin-2-yl hydrazine, 2 1h-pyrazinone, hydrazone, hydrazino pyrazine, pyrazinyl hydrazine, 3-hydrazinopyrazine | |
IVRLZJDPKUSDCF-UHFFFAOYSA-N | |
pyrazin-2-ylhydrazine | |
1487823 | |
98% |
54608-52-5 | |
Yellow | |
MFCD04114555 | |
606630 | |
Slightly soluble in water. | |
C1=CN=C(C=N1)NN | |
110.12 | |
110.12 | |
Solid |
Chemical Identifiers
54608-52-5 | |
110.12 | |
IVRLZJDPKUSDCF-UHFFFAOYSA-N | |
1487823 | |
C1=CN=C(C=N1)NN |
C4H6N4 | |
MFCD04114555 | |
2-hydrazinylpyrazine, 2-hydrazinopyrazine, pyrazin-2-yl-hydrazine, 1-pyrazin-2-yl hydrazine, hydrazinopyrazine, 2-pyrazin-2-yl hydrazine, 2 1h-pyrazinone, hydrazone, hydrazino pyrazine, pyrazinyl hydrazine, 3-hydrazinopyrazine | |
pyrazin-2-ylhydrazine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only