missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydrazinobenzothiazole, 97%, Thermo Scientific Chemicals

$209.60 - $763.53

Chemical Identifiers

CAS 615-21-4
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.214
MDL Number MFCD00041849
InChI Key JYSUYJCLUODSLN-UHFFFAOYSA-N
Synonym 2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine
PubChem CID 11988
IUPAC Name 1,3-benzothiazol-2-ylhydrazine
SMILES C1=CC=C2C(=C1)N=C(S2)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1800609
View Documents Promotion Details
Thermo Scientific Chemicals
A1800609
10 g
Each for $209.60
 
AAA1800618
View Documents Promotion Details
Thermo Scientific Chemicals
A1800618
50 g
Each for $763.53
 
Description

Description

It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.

Solubility
Solubility in hot methanol almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

615-21-4
165.214
JYSUYJCLUODSLN-UHFFFAOYSA-N
11988
C1=CC=C2C(=C1)N=C(S2)NN
C7H7N3S
MFCD00041849
2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine
1,3-benzothiazol-2-ylhydrazine

Specifications

615-21-4
C7H7N3S
10 g
JYSUYJCLUODSLN-UHFFFAOYSA-N
1,3-benzothiazol-2-ylhydrazine
11988
97%
198°C to 202°C
MFCD00041849
2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine
C1=CC=C2C(=C1)N=C(S2)NN
165.214
165.21
2-Hydrazinobenzothiazole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302-H500

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-416-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only