missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydrazinobenzoic Acid Hydrochloride 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 411800050

 View more versions of this product

Catalog No. AC411800050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Hydrazinobenzoic acid hydrochloride
5g
52356-01-1
100.0
Powder
99%
Glass bottle
MFCD00012931
Off-white to beige
99%
98.5
Authentic
≥98.5%
C7H8N2O2·HCl
188.61

Chemical Identifiers

52356-01-1
100.0
C7H8N2O2·HCl
MFCD00012931
≥98.5%
98.5
99%
188.61
2-Hydrazinobenzoic acid hydrochloride, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 257-869-6

TSCA : TSCA