Please login to your online account to display your discounted pricing

2-Hydrazinobenzoic Acid Hydrochloride 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Hydrazinobenzoic acid hydrochloride
Name Note 99%
CAS 52356-01-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O2·HCl
Formula Weight 188.61
MDL Number MFCD00012931
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC411800050

  SDS
thermo fisher scientific
411800050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC411800250

  SDS
thermo fisher scientific
411800250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 188.61
Color Off-white to beige
Quantity 5g
EINECS Number 257-869-6
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Hydrazinobenzoic acid hydrochloride
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H8N2O2·HCl
MDL Number MFCD00012931
CAS 52356-01-1
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.