Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydrazinobenzoic acid hydrochloride, 97%

$37.81 - $37.81

Chemical Identifiers

Chemical Name or Material 2-Hydrazinobenzoic acid hydrochloride
CAS 52356-01-1
Molecular Formula C7H8N2O2·HCl
Formula Weight 188.61
MDL Number MFCD00012931
Beilstein 4011728
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1101506

  SDS
alfa aesar™
L1101506
5g Each for $37.81
Description & Specifications

Specifications

Formula Weight 188.61
Chemical Name or Material 2-Hydrazinobenzoic acid hydrochloride
Health Hazard 1 Warning
Molecular Formula C7H8N2O2·HCl
CAS 52356-01-1
TSCA TSCA
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Beilstein 4011728
Melting Point ≈181° to 183°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00012931
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.