missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydrazinobenzoic acid hydrochloride, 97%

Manufacturer:  Alfa Aesar™ L1101514

 View more versions of this product

Catalog No. AAL1101514



Specifications

Specifications

2-Hydrazinobenzoic acid hydrochloride
25g
C7H9ClN2O2
4011728
ZGNNOFKURIXXRF-UHFFFAOYSA-N
2-hydrazinylbenzoic acid;hydrochloride
104169
97%
~181°C to 183°C
52356-01-1
MFCD00012931
2-hydrazinobenzoic acid hydrochloride, 2-hydrazinylbenzoic acid hydrochloride, benzoic acid, 2-hydrazino-, monohydrochloride, benzoic acid, 2-hydrazino-, hydrochloride, benzoic acid, 2-hydrazinyl-, hydrochloride 1:1, 2-carboxyphenylhydrazine hydrochloride, 2-hydrazinobenzoicacidhydrochloride, 2-carboxyphenylhydrazinium 1+ chloride, pubchem6411, 2-hydrazinobenzoic acid hydrochloride 1:1
C1=CC=C(C(=C1)C(=O)O)NN.Cl
188.611
188.61

Chemical Identifiers

52356-01-1
188.611
ZGNNOFKURIXXRF-UHFFFAOYSA-N
104169
C1=CC=C(C(=C1)C(=O)O)NN.Cl
C7H9ClN2O2
MFCD00012931
2-hydrazinobenzoic acid hydrochloride, 2-hydrazinylbenzoic acid hydrochloride, benzoic acid, 2-hydrazino-, monohydrochloride, benzoic acid, 2-hydrazino-, hydrochloride, benzoic acid, 2-hydrazinyl-, hydrochloride 1:1, 2-carboxyphenylhydrazine hydrochloride, 2-hydrazinobenzoicacidhydrochloride, 2-carboxyphenylhydrazinium 1+ chloride, pubchem6411, 2-hydrazinobenzoic acid hydrochloride 1:1
2-hydrazinylbenzoic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents