missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%

Manufacturer:  Alfa Aesar™ H6128706

 View more versions of this product

Catalog No. AAH6128706



Description

Description

Applications
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

Solubility
Soluble in aqueous acid.

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Hydrazino-4-(trifluoromethyl)pyrimidine
5g
C5H5F3N4
UN2811
XQCAINVUBSAUBJ-UHFFFAOYSA-N
[4-(trifluoromethyl)pyrimidin-2-yl]hydrazine
1268079
97%
99°C to 101°C
197305-97-8
MFCD00115112
1-4-trifluoromethyl pyrimidin-2-yl hydrazine
C1=CN=C(N=C1C(F)(F)F)NN
178.118
178.12

Chemical Identifiers

197305-97-8
178.118
XQCAINVUBSAUBJ-UHFFFAOYSA-N
1268079
C1=CN=C(N=C1C(F)(F)F)NN
C5H5F3N4
MFCD00115112
1-4-trifluoromethyl pyrimidin-2-yl hydrazine
[4-(trifluoromethyl)pyrimidin-2-yl]hydrazine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents