Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hexynal diethyl acetal, 98%

$330.95 - $330.95

Chemical Identifiers

Chemical Name or Material 2-Hexynal diethyl acetal
CAS 18229-78-2
Molecular Formula CH3(CH2)2C≡CCH(OCH2CH3)2
Formula Weight 170.25
MDL Number MFCD00041643
Beilstein 1703624
Synonym 1,1-Diethoxy-2-hexyne
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0609814

  SDS
alfa aesar™
L0609814
25g Each for $330.95
Description & Specifications

Specifications

Formula Weight 170.25
Chemical Name or Material 2-Hexynal diethyl acetal
Assay 98%
Molecular Formula CH3(CH2)2C≡CCH(OCH2CH3)2
MDL Number MFCD00041643
CAS 18229-78-2
Beilstein 1703624
Synonym 1,1-Diethoxy-2-hexyne
Boiling Point 79° to 80°C (8mmHg)
Quantity 25g