Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hexenylsuccinic anhydride, 97%

$78.04 - $78.04

Chemical Identifiers

Chemical Name or Material 2-Hexenylsuccinic anhydride
CAS 39587-79-6
Molecular Formula C10H14O3
Formula Weight 182.22
MDL Number MFCD00061535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1744906

 
alfa aesar™
A1744906
5g Each for $78.04
Description & Specifications

Specifications

Formula Weight 182.22
Chemical Name or Material 2-Hexenylsuccinic anhydride
Assay 97%
Molecular Formula C10H14O3
MDL Number MFCD00061535
CAS 39587-79-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.