missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hexene, cis + trans, tech. 85%

Manufacturer:  Alfa Aesar™ H5347206

 View more versions of this product

Catalog No. AAH5347206



Specifications

Specifications

2-Hexene
−20°C (−4°F)
67°C to 69°C
Cis + Trans
C6H12
MFCD00009473
969179
RYPKRALMXUUNKS-HWKANZROSA-N
(E)-hex-2-ene
639661
85%
0.678
1.395
5g
592-43-8
CH3(CH2)2CH=CHCH3
UN3295
2-hexene z
CCCC=CC
84.162
84.16
Technical

Chemical Identifiers

592-43-8
84.162
RYPKRALMXUUNKS-HWKANZROSA-N
639661
CCCC=CC
C6H12
MFCD00009473
2-hexene z
(E)-hex-2-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 209-755-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents