Learn More
(+/-)-2-Hexanol, 99%
CAS: 626-93-7 | C6H14O | 102.177 g/mol
Supplier: Thermo Scientific Chemicals L0599322
Description
(±)-2-Hexanol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff field. It is used as perfuming agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(+/-)-2-Hexanol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff field. It is used as perfuming agent.
Solubility
Soluble in water (13 g/L at 25°C),
Notes
Incompatible with acid chlorides and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| (+/-)-2-Hexanol | |
| -23°C | |
| 137°C to 139°C | |
| Characteristic | |
| 1.414 | |
| 100 g | |
| 1718996 | |
| Soluble in water (13g/L at 25°C), | |
| CCCCC(C)O | |
| 102.177 | |
| 102.18 |
| 626-93-7 | |
| 0.81 | |
| 45°C (113°F) | |
| C6H14O | |
| MFCD00004585 | |
| UN2282 | |
| 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol | |
| QNVRIHYSUZMSGM-UHFFFAOYSA-N | |
| hexan-2-ol | |
| 12297 | |
| 99% |
Chemical Identifiers
| 626-93-7 | |
| 102.177 | |
| QNVRIHYSUZMSGM-UHFFFAOYSA-N | |
| 12297 | |
| CCCCC(C)O |
| C6H14O | |
| MFCD00004585 | |
| 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol | |
| hexan-2-ol |
Safety and Handling
GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
EINECSNumber : 210-971-4
RTECSNumber : MO8470000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only