missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hexanol, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 156290100

Catalog No. AC156290100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Hexanol
98.5
0.8100g/mL
41°C
98.5% min. (GC)
C6H14O
CH3(CH2)3CH(OH)CH3
10g
0.81
Solubility in water: insoluble.
CCCCC(C)O
102.18
102.18
626-93-7
100.0
139.0°C
Authentic
Glass bottle
1.4125 to 1.4155
MFCD00004585
01, 408
2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol
QNVRIHYSUZMSGM-UHFFFAOYSA-N
hexan-2-ol
12297
99%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 210-971-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only