missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Hexamethyleneimino)ethyl chloride hydrochloride, 98%

Manufacturer:  Alfa Aesar™ L0183314

 View more versions of this product

Catalog No. AAL0183314



Specifications

Specifications

2-(Hexamethyleneimino)ethyl chloride hydrochloride
25g
C8H17Cl2N
UN2811
1-2-chloroethyl azepane hydrochloride, 2-hexamethyleneimino ethyl chloride hydrochloride, 1h-azepine, hydrochloride, 2-chloroethylhexamethylenemine hydrochloride, 1-2-chloroethyl hexamethyleneimine hydrochloride, 2-chloroethyl hexamethylene imine hydrochloride, 2-chloroethyl azaperhydroepine, chloride, pubchem21389, 1-2-chloroethyl azepane hcl, wln: t7ntj a2g &gh
C1CCCN(CC1)CCCl.Cl
198.131
198.14
206°C to 212°C
26487-67-2
MFCD00012842
3680388
ZQDSOUPBYJIPNM-UHFFFAOYSA-N
1-(2-chloroethyl)azepane;hydrochloride
24188283
98%

Chemical Identifiers

26487-67-2
198.131
ZQDSOUPBYJIPNM-UHFFFAOYSA-N
24188283
C1CCCN(CC1)CCCl.Cl
C8H17Cl2N
MFCD00012842
1-2-chloroethyl azepane hydrochloride, 2-hexamethyleneimino ethyl chloride hydrochloride, 1h-azepine, hydrochloride, 2-chloroethylhexamethylenemine hydrochloride, 1-2-chloroethyl hexamethyleneimine hydrochloride, 2-chloroethyl hexamethylene imine hydrochloride, 2-chloroethyl azaperhydroepine, chloride, pubchem21389, 1-2-chloroethyl azepane hcl, wln: t7ntj a2g &gh
1-(2-chloroethyl)azepane;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CM3185000

Documents