Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Heptyne, 98+%

Chemical Identifiers

Chemical Name or Material 2-Heptyne
CAS 1119-65-9
Molecular Formula C7H12
Linear Formula CH3(CH2)3C≡CCH3
Formula Weight 96.17
MDL Number MFCD00039970
UN Number UN3295
Beilstein 1733563
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2152804

  SDS
alfa aesar™
B2152804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2152809

  SDS
alfa aesar™
B2152809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2152818

  SDS
alfa aesar™
B2152818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 96.17
Assay ≥98%
Odor Characteristic
Molecular Formula C7H12
CAS 1119-65-9
UN Number UN3295
Beilstein 1733563
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P261-P243-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Chemical Name or Material 2-Heptyne
Linear Formula CH3(CH2)3C≡CCH3
MDL Number MFCD00039970
Boiling Point 112° to 113°C
Flash Point 11°C (52°F)
Quantity 2g
Safety and Handling

Danger

GHS P Statement
P210-P261-P243-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

GHS H Statement
H225
Highly flammable liquid and vapour.