Please login to your online account to display your discounted pricing

2-Heptyn-1-ol 97%, ACROS Organics™

$25.57 - $25.57

Chemical Identifiers

Chemical Name or Material 2-Heptyn-1-ol
CAS 1002-36-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H12O
Linear Formula CH3(CH2)3C2CH2OH
Formula Weight 112.17
MDL Number MFCD00039541
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387010010

  SDS
thermo fisher scientific
AC387010010
Glass bottle 1g Each for $25.57
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 112.17
Physical Form Liquid
Quantity 1g
Color Clear colorless to yellow
Density 0.88g/mL
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Chemical Name or Material 2-Heptyn-1-ol
Molecular Formula C7H12O
CAS 1002-36-4
MDL Number MFCD00039541
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula CH3(CH2)3C2CH2OH
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.