Learn More
2-Heptylamine, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 103775000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Heptylamine | |
123-82-0 | |
100.0 | |
142.0°C to 144.0°C | |
Authentic | |
Glass bottle | |
1.4175 to 1.4195 | |
MFCD00008101 | |
04,194 | |
15,9982 | |
Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol, ether, petroleum ether,, chloroform and benzene | |
CCCCCC(C)N | |
115.22 | |
115.22 |
≥98% | |
98.5 | |
0.7680g/mL | |
54°C | |
98% min. (GC) | |
C7H17N | |
CH3(CH2)4CH(NH2)CH3 | |
500mL | |
0.768 | |
2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil | |
VSRBKQFNFZQRBM-UHFFFAOYNA-N | |
heptan-2-amine | |
5603 | |
98+% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rin
GHS Signal Word: Danger
EINECSNumber : 204-655-5
RTECSNumber : MQ5425000
TSCA : TSCA
RUO â Research Use Only