missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Heptanone, 99%

Manufacturer:  Alfa Aesar™ A10200AE

 View more versions of this product

Catalog No. AAA10200AE



Description

Description

Applications
2-Heptanone is used as a solvent for resins and lacquers. It is also used as a fragrance in cosmetics and a flavor for foods. It is involved as a reagent in the synthesis of rifamycin derivatives, which exhibit antibacterial activity. Further, it is employed in the preparation of thiazolidin-4-one derivatives as inhibitors of Toxoplasma gondii. In addition to this, it is considered as an alternative local anesthetic to lidocaine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Specifications

Specifications

2-Heptanone
Unpleasant
-35°C
110-43-0
MFCD00009513
1699063
2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
CCCCCC(=O)C
114.188
CHEBI:5672
99%
47°C (116°F)
149°C to 150°C
100mL
C7H14O
UN1110
14,4663
CATSNJVOTSVZJV-UHFFFAOYSA-N
heptan-2-one
8051
114.19

Chemical Identifiers

110-43-0
114.188
CATSNJVOTSVZJV-UHFFFAOYSA-N
8051
heptan-2-one
C7H14O
MFCD00009513
2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
CHEBI:5672
CCCCCC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-AMYL METHYL KETONE

RTECSNumber : MJ5075000

TSCA : TSCA

SDS Documents