Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Furyldimethylsilanol sodium salt

$85.74 - $721.74

Chemical Identifiers

Chemical Name or Material 2-Furyldimethylsilanol sodium salt
CAS 879904-88-8
Molecular Formula C6H9NaO2Si
Formula Weight 164.21
MDL Number MFCD08457663
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3068603

  SDS
alfa aesar™
H3068603
1g Each for $236.57

AAH30686MD

  SDS
alfa aesar™
H30686MD
250mg Each for $85.74

AAH3068606

  SDS
alfa aesar™
H3068606
5g Each for $721.74
Description & Specifications

Specifications

Formula Weight 164.21
MDL Number MFCD08457663
Health Hazard 1 Warning
CAS 879904-88-8
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Furyldimethylsilanol sodium salt
Molecular Formula C6H9NaO2Si
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point ≈92°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.