Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Furylacetone, 99%

$345.05 - $345.05

Chemical Identifiers

Chemical Name or Material 2-Furylacetone
CAS 6975-60-6
Molecular Formula C7H8O2
Formula Weight 124.14
MDL Number MFCD02683084
Synonym 2-Furanyl-2-propanone; 2-Furyl-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2217114

  SDS
alfa aesar™
B2217114
25g Each for $345.05
Description & Specifications

Specifications

Formula Weight 124.14
Chemical Name or Material 2-Furylacetone
TSCA TSCA
Assay 99%
Molecular Formula C7H8O2
MDL Number MFCD02683084
CAS 6975-60-6
Synonym 2-Furanyl-2-propanone; 2-Furyl-2-propanone
Health Hazard 1 Warning
Boiling Point 179° to 180°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Flash Point 65°C (149°F)
Odor Stench
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C

Applications
Suitable for fragrance applications.2-Furylacetone is used as the reactant in the synthesis and transformations of furan derivatives

Solubility
Soluble in alcohol, triacetin.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C