Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH_2

For cellular and molecular biology applications

$108.34 - $396.28

Specifications

Product Type 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH_2
Molecular Weight 777.97
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence 2-Furoyl-LIGRLO-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66850LB0

 
alfa aesar™
J66850LB0
5mg Each for $396.28

AAJ66850MCR

 
alfa aesar™
J66850MCR
1mg Each for $108.34
Description & Specifications

Specifications

Product Type 2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH_2
Molecular Weight 777.97
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence 2-Furoyl-LIGRLO-NH2
Common Name Protease-Activated Receptor 2 (PAR2) Agonist
Regulatory Status RUO
Purification Method HPLC