missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Furoyl chloride, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 165385000

 View more versions of this product

Catalog No. AC165385000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Furoyl chloride
85°C
Glass bottle
1.32
Yellow to Brown
500mL
527-69-5
100.0
C5H3ClO2
18, 276
2-furancarbonyl chloride
OFTKFKYVSBNYEC-UHFFFAOYSA-N
furan-2-carbonyl chloride
68242
Liquid
1.3200g/mL
Authentic
1.5300 to 1.532
173°C to 174°C
-2°C
98+%
98.0
98% min. (GC)
MFCD00003228
15, 4339
Solubility in water: decomposes. Other solubilities: soluble in ether,toluene,methylene chloride,,dichloroethane,chlorobenzene,ethyl acetate,thf,and chloroform,decomposes in alcohol
C1=COC(=C1)C(=O)Cl
130.53
130.53
98+%

Chemical Identifiers

527-69-5
130.53
OFTKFKYVSBNYEC-UHFFFAOYSA-N
68242
C1=COC(=C1)C(=O)Cl
C5H3ClO2
MFCD00003228
2-furancarbonyl chloride
furan-2-carbonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 208-422-9

TSCA : TSCA

SDS Documents