missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Furonitrile, 99%, ACROS Organics™

Manufacturer:  Acros Organics 170430250

 View more versions of this product

Catalog No. AC170430250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Furonitrile
617-90-3
100.0
99%
Glass bottle
MFCD00003223
18, 278
Product may darken on storage
C1=COC(=C1)C#N
93.085
1.0600g/mL
Liquid
Yellow through Orange
35°C
99%
98.5
Authentic
99%
C5H3NO
1.4780 to 1.4800
2-Cyanofuran
YXDXXGXWFJCXEB-UHFFFAOYSA-N
furan-2-carbonitrile
69245
93.08
146.0°C to 148.0°C
25g

Chemical Identifiers

617-90-3
93.085
YXDXXGXWFJCXEB-UHFFFAOYSA-N
69245
C5H3NO
MFCD00003223
2-Cyanofuran
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 210-537-4

TSCA : TSCA

SDS Documents