Learn More
2-Furonitrile, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 170430050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Furonitrile | |
617-90-3 | |
100.0 | |
146.0°C to 148.0°C | |
Authentic | |
Glass bottle | |
1.4780 to 1.4800 | |
5g | |
1.06 | |
Solubility in water: slightly. Other solubilities: soluble in most common organic solvents | |
N#CC1=CC=CO1 | |
93.09 | |
93.08 |
99% | |
98.5 | |
1.0600g/mL | |
35°C | |
98.5% min. (GC) | |
C5H3NO | |
MFCD00003223 | |
18, 278 | |
2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile | |
YXDXXGXWFJCXEB-UHFFFAOYSA-N | |
furan-2-carbonitrile | |
69245 | |
99% |
Chemical Identifiers
617-90-3 | |
93.09 | |
YXDXXGXWFJCXEB-UHFFFAOYSA-N | |
69245 | |
N#CC1=CC=CO1 |
C5H3NO | |
MFCD00003223 | |
2-furonitrile, 2-cyanofuran, 2-furancarbonitrile, 2-furyl cyanide, unii-2lrk86h722, .alpha.-furyl cyanide, pubchem6961, acmc-209mwb, 2-cyanofuran;2-furancarbonitrile | |
furan-2-carbonitrile |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 210-537-4
TSCA : TSCA
Recommended Storage : Product may darken on storage
RUO â Research Use Only