missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Furfurylthio)ethylamine, TCI America™

Manufacturer:  TCI America F037825G

 View more versions of this product

Catalog No. F037825G



Specifications

Specifications

2-(Furfurylthio)ethylamine
Yellow
36415-21-1
MFCD01075029
VSPDYEHKAMKDNW-UHFFFAOYSA-N
2-(furan-2-ylmethylsulfanyl)ethanamine
118970
Clear Liquid at 20°C
100°C
25g
C7H11NOS
(2-Aminoethylthio)methylfuran
C1=COC(=C1)CSCCN
157.231
157.23
≥98.0% (GC,T)

Chemical Identifiers

36415-21-1
157.231
VSPDYEHKAMKDNW-UHFFFAOYSA-N
118970
C1=COC(=C1)CSCCN
C7H11NOS
MFCD01075029
(2-Aminoethylthio)methylfuran
2-(furan-2-ylmethylsulfanyl)ethanamine
Safety and Handling

Safety and Handling

TSCA : No

Documents