Please login to your online account to display your discounted pricing

(2-Furanylmethylene)malononitrile, 98+%, Acros Organics

Chemical Identifiers

Chemical Name or Material (2-Furanylmethylene)malononitrile
Assay Percent Range 98+%
Molecular Formula C8H4N2O
Formula Weight 114.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC374640050

  SDS
thermo fisher scientific
AC374640050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC374640250

  SDS
thermo fisher scientific
AC374640250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.13
Quantity 5g
Chemical Name or Material (2-Furanylmethylene)malononitrile
Molecular Formula C8H4N2O
CAS 3237-22-7
Assay Percent Range 98+%
Packaging Glass bottle