missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Furaldehyde, ACS, 98% min

Manufacturer:  Alfa Aesar™ 3130530*

 View more versions of this product

Catalog No. AA3130530



Specifications

Specifications

2-Furaldehyde
60° (140°F)
1.5261
162°C
-38°C
98-01-1
MFCD00003229
105755
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
C1=COC(=C1)C=O
96.085
CHEBI:34768
Liquid
ACS
1.159g/mL
Almond-like
Air Sensitive
Yellow to Brown
250g
C5H4O2
UN1199
14,4304
HYBBIBNJHNGZAN-UHFFFAOYSA-N
furan-2-carbaldehyde
7362
96.09
≤98%

Chemical Identifiers

98-01-1
96.085
HYBBIBNJHNGZAN-UHFFFAOYSA-N
7362
furan-2-carbaldehyde
C5H4O2
MFCD00003229
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
CHEBI:34768
C1=COC(=C1)C=O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H226-H351-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: FURALDEHYDES

RTECSNumber : LT7000000

TSCA : TSCA

SDS Documents