Learn More
2-Furaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1616730
Description
2-Furaldehyde is used in the manufacture of furfural-phenol plastics such as Durite; in solvent refining of petroleum oils and in the preparation of pyromucic acid. It is used as a solvent for nitrated cotton, cellulose acetate and gums; for accelerating vulcanization; in the synthesis of furan derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Furaldehyde is used in the manufacture of furfural-phenol plastics such as Durite; in solvent refining of petroleum oils and in the preparation of pyromucic acid. It is used as a solvent for nitrated cotton, cellulose acetate and gums; for accelerating vulcanization; in the synthesis of furan derivatives.
Solubility
Soluble in water (83 g/L).
Notes
Air sensitive. Incompatible with oxidizing agents and acids.
Specifications
2-Furaldehyde | |
-38°C | |
162°C | |
Almond-like | |
1.5261 | |
250 g | |
105755 | |
14,4304 | |
Soluble in water (83g/L). | |
O=CC1=CC=CO1 | |
96.09 | |
CHEBI:34768 | |
98% |
98-01-1 | |
1.159 | |
60°C (140°F) | |
C5H4O2 | |
MFCD00003229 | |
UN1199 | |
Air Sensitive | |
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural | |
HYBBIBNJHNGZAN-UHFFFAOYSA-N | |
furan-2-carbaldehyde | |
7362 | |
96.09 |
Chemical Identifiers
98-01-1 | |
96.09 | |
HYBBIBNJHNGZAN-UHFFFAOYSA-N | |
7362 | |
furan-2-carbaldehyde |
C5H4O2 | |
MFCD00003229 | |
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural | |
CHEBI:34768 | |
O=CC1=CC=CO1 |
Safety and Handling
GHS H Statement
H301-H331-H226-H351-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H312-H315-H319-H330-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: FURALDEHYDES
EINECSNumber : 202-627-7
RTECSNumber : LT7000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only