missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Formylthiophene-4-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H5329806

 View more versions of this product

Catalog No. AAH5329806



Description

Description

Applications
2-Formylthiophene-4-boronic acid is used as intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat. Refrigerate.
Specifications

Specifications

2-Formylthiophene-4-boronic acid
5g
C5H5BO3S
2-formylthiophene-4-boronic acid, 5-formylthiophen-3-yl boronic acid, 5-formylthiophene-3-boronic acid, 5-formyl-3-thienyl boronic acid, 4-borono-2-formylthiophene, 5-formylthiophen-3-boronic acid, 5-formyl-3-thienylboronic acid, 5-formyl-3-thiopheneboronic acid, 4-boronothiophene-2-carboxaldehyde, 2-formyl-4-thiopheneboronic acid
B(C1=CSC(=C1)C=O)(O)O
155.962
156.97
113°C
175592-59-3
MFCD03002365
ZCUFJAORCNFWOF-UHFFFAOYSA-N
(5-formylthiophen-3-yl)boronic acid
11084115
98%

Chemical Identifiers

175592-59-3
155.962
ZCUFJAORCNFWOF-UHFFFAOYSA-N
11084115
B(C1=CSC(=C1)C=O)(O)O
C5H5BO3S
MFCD03002365
2-formylthiophene-4-boronic acid, 5-formylthiophen-3-yl boronic acid, 5-formylthiophene-3-boronic acid, 5-formyl-3-thienyl boronic acid, 4-borono-2-formylthiophene, 5-formylthiophen-3-boronic acid, 5-formyl-3-thienylboronic acid, 5-formyl-3-thiopheneboronic acid, 4-boronothiophene-2-carboxaldehyde, 2-formyl-4-thiopheneboronic acid
(5-formylthiophen-3-yl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

SDS Documents