missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Formylphenoxyacetic acid, 98%

Manufacturer:  Alfa Aesar™ A1460514

 View more versions of this product

Catalog No. AAA1460514



Specifications

Specifications

2-Formylphenoxyacetic acid
25g
C9H8O4
2096797
ANWMNLAAFDCKMT-UHFFFAOYSA-N
2-(2-formylphenoxy)acetic acid
46533
98%
131°C to 134°C
6280-80-4
MFCD00003315
2-formylphenoxyacetic acid, 2-2-formylphenoxy acetic acid, 2-formylphenoxy acetic acid, o-formylphenoxyacetic acid, acetic acid, 2-formylphenoxy, 2-carboxymethoxybenzaldehyde, ortho-formylphenoxyacetic acid, acetic acid, o-formylphenoxy, acetic acid, 2-2-formylphenoxy, 2-formylphenoxyaceticacid
C1=CC=C(C(=C1)C=O)OCC(=O)O
180.159
180.16

Chemical Identifiers

6280-80-4
180.159
ANWMNLAAFDCKMT-UHFFFAOYSA-N
46533
C1=CC=C(C(=C1)C=O)OCC(=O)O
C9H8O4
MFCD00003315
2-formylphenoxyacetic acid, 2-2-formylphenoxy acetic acid, 2-formylphenoxy acetic acid, o-formylphenoxyacetic acid, acetic acid, 2-formylphenoxy, 2-carboxymethoxybenzaldehyde, ortho-formylphenoxyacetic acid, acetic acid, o-formylphenoxy, acetic acid, 2-2-formylphenoxy, 2-formylphenoxyaceticacid
2-(2-formylphenoxy)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents