Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Formylphenoxyacetic acid, 98%

$48.03 - $137.91

Chemical Identifiers

Chemical Name or Material 2-Formylphenoxyacetic acid
CAS 6280-80-4
Molecular Formula C9H8O4
Formula Weight 180.16
MDL Number MFCD00003315
Beilstein 2096797
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1460514

 
alfa aesar™
A1460514
25g Each for $48.03

AAA1460522

 
alfa aesar™
A1460522
100g Each for $137.91
Description & Specifications

Specifications

Formula Weight 180.16
Chemical Name or Material 2-Formylphenoxyacetic acid
TSCA TSCA
Assay 98%
Molecular Formula C9H8O4
MDL Number MFCD00003315
CAS 6280-80-4
Beilstein 2096797
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 131° to 134°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.