missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Formylbenzenesulfonic acid sodium salt hydrate, tech. 90%

$41.92 - $111.99

Chemical Identifiers

CAS 305808-14-4
Molecular Formula C7H8NaO5S
Molecular Weight (g/mol) 227.186
MDL Number MFCD00007478
InChI Key LYIJOFWETPXEGE-UHFFFAOYSA-N
Synonym sodium 2-formylbenzenesulfonic acid hydrate
PubChem CID 87238610
IUPAC Name 2-formylbenzenesulfonic acid;sodium;hydrate
SMILES C1=CC=C(C(=C1)C=O)S(=O)(=O)O.O.[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1973618
View Documents
Alfa Aesar™
A1973618
50g
Each for $41.92
 
AAA1973630
View Documents
Alfa Aesar™
A1973630
250g
Each for $111.99
 
Description

Description

Specifications

Chemical Identifiers

305808-14-4
227.186
LYIJOFWETPXEGE-UHFFFAOYSA-N
87238610
C1=CC=C(C(=C1)C=O)S(=O)(=O)O.O.[Na]
C7H8NaO5S
MFCD00007478
sodium 2-formylbenzenesulfonic acid hydrate
2-formylbenzenesulfonic acid;sodium;hydrate

Specifications

Air Sensitive
305808-14-4
MFCD00007478
LYIJOFWETPXEGE-UHFFFAOYSA-N
2-formylbenzenesulfonic acid;sodium;hydrate
87238610
90%
2-Formylbenzenesulfonic acid sodium salt hydrate
50g
C7H8NaO5S
sodium 2-formylbenzenesulfonic acid hydrate
C1=CC=C(C(=C1)C=O)S(=O)(=O)O.O.[Na]
227.186
208.17 (Anhydrous)
Technical
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-053-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS