missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC219320025

 View more versions of this product

Catalog No. AC219320025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

sodium chloride: < 3%
White to pale yellow
tech., 90%
88.0
Authentic
Technical Grade
≥88%
C7H5NaO4S
208.17
11, 324
2-Formylbenzenesulfonic acid sodium salt
2.5kg
1008-72-6
100.0
Plastic bottle
Powder
90%
HCOC6H4SO3Na
MFCD00007478
Benzaldehyde-2-sulfonic acid, sodium salt

Chemical Identifiers

1008-72-6
100.0
C7H5NaO4S
208.17
Benzaldehyde-2-sulfonic acid, sodium salt
≥88%
88.0
90%
HCOC6H4SO3Na
MFCD00007478
2-Formylbenzenesulfonic acid sodium salt, tech., 90%
Technical Grade

Safety and Handling

SDS & Safety Documents

AIR SENSITIVE

EINECSNumber : 213-758-4

TSCA : TSCA