Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Formylbenzeneboronic acid pinacol ester, 97%

$80.96 - $240.12

Chemical Identifiers

Chemical Name or Material 2-Formylbenzeneboronic acid pinacol ester
CAS 380151-85-9
Molecular Formula C13H17BO3
Formula Weight 232.08
MDL Number MFCD07363841
Synonym 2-Formylphenylboronic acid pinacol ester; 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2884603

 
alfa aesar™
H2884603
1g Each for $80.96

AAH2884606

 
alfa aesar™
H2884606
5g Each for $240.12
Description & Specifications

Specifications

Formula Weight 232.08
Chemical Name or Material 2-Formylbenzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C13H17BO3
MDL Number MFCD07363841
CAS 380151-85-9
Synonym 2-Formylphenylboronic acid pinacol ester; 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.