missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Fmoc-amino)benzoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6345914
Description
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
Solubility
Soluble in water or 1% acetic acid.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
2-(Fmoc-amino)benzoic acid | |
216°C to 218°C (decomposition) | |
C22H17NO4 | |
25 g | |
fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid | |
CNAVPEPPAVHHKN-UHFFFAOYSA-N | |
2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid | |
978386 | |
98% |
150256-42-1 | |
White | |
MFCD00235883 | |
7659726 | |
Soluble in water or 1% acetic acid. | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O | |
359.381 | |
359.37 | |
Powder |
Chemical Identifiers
150256-42-1 | |
359.381 | |
CNAVPEPPAVHHKN-UHFFFAOYSA-N | |
978386 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O |
C22H17NO4 | |
MFCD00235883 | |
fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid | |
2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only