Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Fmoc-amino)-4-chlorobenzoic acid, 95%

$94.93 - $94.93

Chemical Identifiers

Chemical Name or Material 2-(Fmoc-amino)-4-chlorobenzoic acid
CAS 332121-92-3
Molecular Formula C22H16ClNO4
Formula Weight 393.83
MDL Number MFCD02682157
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-chloroanthranilic acid; N-Fmoc-4-chloroanthranilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1980903

 
alfa aesar™
L1980903
1g Each for $94.93
Description & Specifications

Specifications

Formula Weight 393.83
Chemical Name or Material 2-(Fmoc-amino)-4-chlorobenzoic acid
CAS 332121-92-3
MDL Number MFCD02682157
Health Hazard 1 Warning
Molecular Formula C22H16ClNO4
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Melting Point 218° to 220°C
Quantity 1g
Assay 95%
Synonym N-(9-Fluorenylmethoxycarbonyl)-4-chloroanthranilic acid; N-Fmoc-4-chloroanthranilic acid
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.