Please login to your online account to display your discounted pricing

2-Fluorothiophenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluorothiophenol
Name Note 97%
CAS 2557-78-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H5FS
Linear Formula FC6H4SH
Formula Weight 128.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369880010

  SDS
thermo fisher scientific
AC369880010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 128.17
Quantity 1g
Color Clear colorless to light yellow
Density 1.2030g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Fluorothiophenol
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6H5FS
MDL Number MFCD00041419
Synonym 2-Fluorobenzenethiol
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Packaging Glass bottle
Refractive Index 1.5530 to 1.5570 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula FC6H4SH
CAS 2557-78-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.