Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoroquinoline-3-boronic acid, 97%

$127.76 - $1,431.53

Chemical Identifiers

Chemical Name or Material 2-Fluoroquinoline-3-boronic acid
CAS 745784-10-5
Molecular Formula C9H7BFNO2
Formula Weight 190.97
MDL Number MFCD05664131
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2034103

 
alfa aesar™
L2034103
1g Each for $367.89

AAL2034106

 
alfa aesar™
L2034106
5g Each for $1,431.53

AAL20341MD

 
alfa aesar™
L20341MD
250mg Each for $127.76
Description & Specifications

Specifications

Formula Weight 190.97
MDL Number MFCD05664131
Chemical Name or Material 2-Fluoroquinoline-3-boronic acid
Assay 97%
CAS 745784-10-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 89° to 94°C
Molecular Formula C9H7BFNO2
Health Hazard 1 Warning
Quantity 1g

Applications
2-Fluoroquinoline-3-boronic acid acts as a pharmaceutical intermediate. It is also useful as a fluorinated building block in synthetic chemistry.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.