missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoropyridine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119630500

 View more versions of this product

Catalog No. AC119630500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Fluoropyridine
24°C
Glass bottle
1.12
Colorless to Yellow
372-48-5
100.0
C5H4FN
MFCD00006224
2-fluoranylpyridine, 2-fluoro-pyridine, 2-fluoro-pyridinium, 2-fluorpyridin, 2fluoropyridine, ccris 1725, fluoropyridine, o-fluoropyridine, pyridine, 2-fluoro, pyridine, fluoro
C1=CC=NC(=C1)F
97.09
97.09
98%
1.1200g/mL
Authentic
1.4670 to 1.4690
126.0°C (753.0 mmHg)
50mL
97.5
98%
(C5H4N)F
20, I, 80
MTAODLNXWYIKSO-UHFFFAOYSA-N
2-fluoropyridine
9746
Liquid

Chemical Identifiers

372-48-5
97.09
MTAODLNXWYIKSO-UHFFFAOYSA-N
9746
C1=CC=NC(=C1)F
C5H4FN
MFCD00006224
2-fluoranylpyridine, 2-fluoro-pyridine, 2-fluoro-pyridinium, 2-fluorpyridin, 2fluoropyridine, ccris 1725, fluoropyridine, o-fluoropyridine, pyridine, 2-fluoro, pyridine, fluoro
2-fluoropyridine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-757-5

SDS Documents