missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Fluoro-3-pyridinylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 351019-18-6
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.91
InChI Key OJBYZWHAPXIJID-UHFFFAOYSA-N
Synonym 2-fluoro-5-pyridineboronic acid, 2-fluoro-5-pyridylboronic acid, 2-fluoropyridin-5-boronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridine-5-boronic acid, 6-fluoro-3-pyridineboronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoropyridine-3-boronic acid
PubChem CID 2783397
IUPAC Name (6-fluoropyridin-3-yl)boronic acid
SMILES B(C1=CN=C(C=C1)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC397160010 SDS View Documents Thermo Fisher Scientific
AC397160010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397160050 SDS View Documents Thermo Fisher Scientific
AC397160050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

351019-18-6
140.91
2-fluoro-5-pyridineboronic acid, 2-fluoro-5-pyridylboronic acid, 2-fluoropyridin-5-boronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridine-5-boronic acid, 6-fluoro-3-pyridineboronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoropyridine-3-boronic acid
(6-fluoropyridin-3-yl)boronic acid
C5H5BFNO2
OJBYZWHAPXIJID-UHFFFAOYSA-N
2783397
B(C1=CN=C(C=C1)F)(O)O

Specifications

Glass bottle
1g
96.0
97%
2-fluoro-5-pyridineboronic acid, 2-fluoro-5-pyridylboronic acid, 2-fluoropyridin-5-boronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridine-5-boronic acid, 6-fluoro-3-pyridineboronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoropyridine-3-boronic acid
B(C1=CN=C(C=C1)F)(O)O
140.91
140.91
6-Fluoro-3-pyridinylboronic acid
172.0°C to 176.0°C
351019-18-6
100.0
C5H5BFNO2
OJBYZWHAPXIJID-UHFFFAOYSA-N
(6-fluoropyridin-3-yl)boronic acid
2783397
97%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning