missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

$478.85 - $1208.53

Chemical Identifiers

CAS 458532-86-0
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.05
MDL Number MFCD06798253
InChI Key PCLMNCBIXQQRMB-UHFFFAOYSA-N
Synonym 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12060164
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2717903
View Documents Promotion Details
Thermo Scientific Chemicals
H2717903
1 g
Each for $478.85
 
AAH2717906
View Documents Promotion Details
Thermo Scientific Chemicals
H2717906
5 g
Each for $1,208.53
 
Description

Description

Used in pharmaceuticals, agrochemicals, and high-tech materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceuticals, agrochemicals, and high-tech materials.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

458532-86-0
223.05
PCLMNCBIXQQRMB-UHFFFAOYSA-N
12060164
CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1
C11H15BFNO2
MFCD06798253
2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Specifications

458532-86-0
C11H15BFNO2
1 g
PCLMNCBIXQQRMB-UHFFFAOYSA-N
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
12060164
95%
48°C to 52°C
MFCD06798253
2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1
223.05
223.06
2-Fluoropyridine-4-boronic acid pinacol ester
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only