missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoropyridine-3-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 393-55-5
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.1
InChI Key LLLVHTWJGWNRBD-UHFFFAOYSA-N
Synonym 2-fluoro nicotinic acid
PubChem CID 96089
IUPAC Name 2-fluoropyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC436820010 SDS View Documents Thermo Fisher Scientific
AC436820010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436820050 SDS View Documents Thermo Fisher Scientific
AC436820050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

393-55-5
141.1
2-fluoro nicotinic acid
2-fluoropyridine-3-carboxylic acid
C6H4FNO2
LLLVHTWJGWNRBD-UHFFFAOYSA-N
96089
C1=CC(=C(N=C1)F)C(=O)O

Specifications

Authentic
White to Yellow
1g
96.0
96% min. (HPLC)
2-fluoro nicotinic acid
C1=CC(=C(N=C1)F)C(=O)O
141.1
141.1
97%
Glass Bottle
150.8°C to 151.4°C
393-55-5
100.0
C6H4FNO2
LLLVHTWJGWNRBD-UHFFFAOYSA-N
2-fluoropyridine-3-carboxylic acid
96089
Powder or Crystalline Powder
2-Fluoropyridine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 206-888-8