missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoropyridine-3-carboxylic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436820050
Specifications
2-Fluoropyridine-3-carboxylic acid | |
Glass bottle | |
150.8°C to 151.4°C | |
393-55-5 | |
100.0 | |
C6H4FNO2 | |
LLLVHTWJGWNRBD-UHFFFAOYSA-N | |
2-fluoropyridine-3-carboxylic acid | |
96089 | |
Powder or Crystalline Powder |
Authentic | |
White to Yellow | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-fluoro nicotinic acid, 2-fluoro-3-carboxypyridine, 2-fluoro-3-pyridinecarboxylic acid, 2-fluoro-3-pyridinecarboxylic acid 2-fluoronicotinic acid, 2-fluoro-nicotinic acid, 2-fluoronicotinic acid, 2-fluoronicotinicacid, 3-pyridinecarboxylic acid, 2-fluoro, fluoronicotinic acid, pubchem1120 | |
C1=CC(=C(N=C1)F)C(=O)O | |
141.1 | |
141.1 | |
97% |
Chemical Identifiers
393-55-5 | |
141.1 | |
2-fluoro nicotinic acid, 2-fluoro-3-carboxypyridine, 2-fluoro-3-pyridinecarboxylic acid, 2-fluoro-3-pyridinecarboxylic acid 2-fluoronicotinic acid, 2-fluoro-nicotinic acid, 2-fluoronicotinic acid, 2-fluoronicotinicacid, 3-pyridinecarboxylic acid, 2-fluoro, fluoronicotinic acid, pubchem1120 | |
2-fluoropyridine-3-carboxylic acid |
C6H4FNO2 | |
LLLVHTWJGWNRBD-UHFFFAOYSA-N | |
96089 | |
C1=CC(=C(N=C1)F)C(=O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
EINECSNumber : 206-888-8