Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoropyridine-3-boronic acid pinacol ester, 95%

$158.22 - $354.09

Chemical Identifiers

Chemical Name or Material 2-Fluoropyridine-3-boronic acid pinacol ester
CAS 452972-14-4
Molecular Formula C11H15BFNO2
Formula Weight 223.06
MDL Number MFCD06798245
Synonym 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2784503

  SDS
alfa aesar™
H2784503
1g Each for $354.09

AAH27845MD

  SDS
alfa aesar™
H27845MD
250mg Each for $158.22
Description & Specifications

Specifications

Formula Weight 223.06
Chemical Name or Material 2-Fluoropyridine-3-boronic acid pinacol ester
Assay 95%
Molecular Formula C11H15BFNO2
MDL Number MFCD06798245
CAS 452972-14-4
Synonym 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 42° to 45°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.