Please login to your online account to display your discounted pricing

2-Fluoropyridine-3-boronic acid 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393510010

 View more versions of this product

Catalog No. AC393510010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 140.91
Quantity 1g
Color White
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
MDL Number MFCD03411557
Packaging Glass bottle
Chemical Name or Material 2-Fluoropyridine-3-boronic acid
Molecular Formula C5H5BFNO2
CAS 174669-73-9
Assay Percent Range 97%
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.

Specifications

Chemical Name or Material 2-Fluoropyridine-3-boronic acid
CAS 174669-73-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H5BFNO2
Formula Weight 140.91
MDL Number MFCD03411557
Accessories

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.

SDS & Safety Documents