Please login to your online account to display your discounted pricing

2-Fluoropyridine-3-boronic acid 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoropyridine-3-boronic acid
CAS 174669-73-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H5BFNO2
Formula Weight 140.91
MDL Number MFCD03411557
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393510010

  SDS
thermo fisher scientific
AC393510010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 140.91
Quantity 1g
Color White
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD03411557
Chemical Name or Material 2-Fluoropyridine-3-boronic acid
Assay Percent Range 97%
Molecular Formula C5H5BFNO2
CAS 174669-73-9
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Safety and Handling

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.