Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoropyridine-3-boronic acid, 97%

$63.73 - $290.81

Chemical Identifiers

Chemical Name or Material 2-Fluoropyridine-3-boronic acid
CAS 174669-73-9
Molecular Formula C5H5BFNO2
Formula Weight 140.91
MDL Number MFCD03411557
Beilstein 9119935
Synonym 2-Fluoro-3-pyridylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2010803

  SDS
alfa aesar™
L2010803
1g Each for $63.73

AAL2010806

  SDS
alfa aesar™
L2010806
5g Each for $290.81
Description & Specifications

Specifications

Formula Weight 140.91
Synonym 2-Fluoro-3-pyridylboronic acid
Quantity 1g
CAS 174669-73-9
MDL Number MFCD03411557
Chemical Name or Material 2-Fluoropyridine-3-boronic acid
Molecular Formula C5H5BFNO2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Beilstein 9119935
Melting Point 172°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.