Please login to your online account to display your discounted pricing

2-Fluorophenylhydrazine hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluorophenylhydrazine hydrochloride
CAS 2924-15-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H7FN2·ClH
Linear Formula FC6H4NHNH2·HCl
Formula Weight 162.59
MDL Number MFCD00012927
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC167050010

  SDS
thermo fisher scientific
167050010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 162.59
Quantity 1g
Color Beige to brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H7FN2·ClH
Linear Formula FC6H4NHNH2·HCl
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2-Fluorophenylhydrazine hydrochloride
Infrared Spectrum Authentic
MDL Number MFCD00012927
CAS 2924-15-4
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
Suspected of causing cancer.