missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluorophenylacetylene, 97%

$53.51 - $178.89

Chemical Identifiers

CAS 766-49-4
Molecular Formula C8H5F
Molecular Weight (g/mol) 120.126
MDL Number MFCD00799540
InChI Key YFPQIXUNBPQKQR-UHFFFAOYSA-N
Synonym 2-fluorophenylacetylene, 2'-fluorophenyl acetylene, 1-ethynyl-2-fluoro-benzene, 2'-fluorophenylacetylene, 2-fluorophenylethyne, o-fluorophenylacetylene, benzene, ethynylfluoro, 2-fluoro-phenylacetylene, 2-fluorophenyl-ethyne, 2-fluorophenyl acetylene
PubChem CID 643358
IUPAC Name 1-ethynyl-2-fluorobenzene
SMILES C#CC1=CC=CC=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60596MD View Documents Alfa Aesar™
H60596MD
250mg Each for $53.51
 
AAH6059603 View Documents Alfa Aesar™
H6059603
1g Each for $178.89
 
Specifications

Chemical Identifiers

766-49-4
120.126
YFPQIXUNBPQKQR-UHFFFAOYSA-N
643358
C#CC1=CC=CC=C1F
C8H5F
MFCD00799540
2-fluorophenylacetylene, 2'-fluorophenyl acetylene, 1-ethynyl-2-fluoro-benzene, 2'-fluorophenylacetylene, 2-fluorophenylethyne, o-fluorophenylacetylene, benzene, ethynylfluoro, 2-fluoro-phenylacetylene, 2-fluorophenyl-ethyne, 2-fluorophenyl acetylene
1-ethynyl-2-fluorobenzene

Specifications

37°C (99°F)
250mg
C8H5F
UN1993
YFPQIXUNBPQKQR-UHFFFAOYSA-N
1-ethynyl-2-fluorobenzene
643358
97%
150°C
766-49-4
MFCD00799540
2-fluorophenylacetylene, 2'-fluorophenyl acetylene, 1-ethynyl-2-fluoro-benzene, 2'-fluorophenylacetylene, 2-fluorophenylethyne, o-fluorophenylacetylene, benzene, ethynylfluoro, 2-fluoro-phenylacetylene, 2-fluorophenyl-ethyne, 2-fluorophenyl acetylene
C#CC1=CC=CC=C1F
120.126
120.12
2-Fluorophenylacetylene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.