missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorophenylacetonitrile, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1161206

 View more versions of this product

Catalog No. AAA1161206



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluorophenylacetonitrile
1.061
>100°C (212°F)
1.501
5 g
1862361
DAVJMKMVLKOQQC-UHFFFAOYSA-N
2-(2-fluorophenyl)acetonitrile
67592
97%
326-62-5
235°C
C8H6FN
MFCD00001897
UN3276
2-fluorophenylacetonitrile, 2-fluorobenzyl cyanide, 2-2-fluorophenyl acetonitrile, benzeneacetonitrile, 2-fluoro, o-fluorophenylacetonitrile, o-fluorobenzyl cyanide, 2-fluorophenyl acetonitrile, acetonitrile, o-fluorophenyl, 2-2-fluorophenyl ethanenitrile, 2-fluorobenzeneacetonitrile
FC1=CC=CC=C1CC#N
135.14
135.14

Chemical Identifiers

326-62-5
135.14
DAVJMKMVLKOQQC-UHFFFAOYSA-N
67592
FC1=CC=CC=C1CC#N
C8H6FN
MFCD00001897
2-fluorophenylacetonitrile, 2-fluorobenzyl cyanide, 2-2-fluorophenyl acetonitrile, benzeneacetonitrile, 2-fluoro, o-fluorophenylacetonitrile, o-fluorobenzyl cyanide, 2-fluorophenyl acetonitrile, acetonitrile, o-fluorophenyl, 2-2-fluorophenyl ethanenitrile, 2-fluorobenzeneacetonitrile
2-(2-fluorophenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H226-H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 206-313-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only